missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Interferon alpha 2b Protein is fully biologically active when compared to standard. The specific activity determined by an anti-viral assay is no less than 1.6 x 10^8 IU/mg.
Specifications
Specifications
| Accession Number | P01563, P01563, P01563 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in PBS, pH 7.4, with 0.02 % Tween-20. |
| Gene ID (Entrez) | 3440 |
| Molecular Weight (g/mol) | 19.3 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of IFN-alpha 2 as determined by LAL method. |
| Gene Alias | alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?