missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IFN-alpha 10/IFNA10 Protein

Product Code. 18789196
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18789196

Brand: R&D Systems™ 11016IF010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is 1.00-30.0 pg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01566.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3446
Molecular Weight (g/mol) MolecularWeight-observed: 18-22 kDa, under reducing conditions., MolecularWeight-theroretical: 19 kDa
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human IFN-alpha 10/IFNA10 protein Cys24-Asp189
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IFNA10, IFNalpha C, IFN-alpha C, IFN-alpha-10, IFN-alphaC, interferon alpha-10, Interferon alpha-6 L, Interferon alpha-C, interferon, alpha 10, LeIF C, MGC119878, MGC119879
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IFN-alpha C/IFN-alpha 10
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.