missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IFN-alpha 1 Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
Recombinant bioactive protein containing 166 amino acids for Human IFN-alpha D The Recombinant Human IFN-alpha 1 Protein is derived from E. coli. The Recombinant Human IFN-alpha 1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NM_024013 |
| Formulation | Sterile filtered and lyophilized with no additives. |
| Gene ID (Entrez) | 3439 |
| Name | IFN-alpha D Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Reconstitution | Reconstitute in H2O to a concentration of 0.1 - 1.0 mgml. The solution can then be diluted into other aqueous buffers and stored at 4C for 1 week or -20C for long-term storage. |
| Purity or Quality Grade | >95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction