missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ICOS (aa 21-140) mFc Chimera Protein, CF
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. Recombinant Human ICOS mFc Chimera (Catalog # 11423-CS) binds to Recombinant Human B7-H2 Fc Chimera Protein (Catalog # 165-B7 ) with a ED50 of 1.50-15.0 ng/mL.
Specifications
Specifications
| Accession Number | Q9Y6W8.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 29851 |
| Molecular Weight (g/mol) | M.W. (Observed): 54-60 kDa, under reducing conditions., M.W. (Theroretical): 41 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human ICOS protein Human ICOS (Glu21-Lys140) (N-terminus) Accession # Q9Y6W8.1 IEGRMD Mouse IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction