missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ICAM-2/CD102 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 803-I2-200
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human ICAM-2/CD102 Fc Chimera Protein is derived from NS0. The Recombinant Human ICAM-2/CD102 Fc Chimera Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol. | |
50 kDa (monomer) | |
200μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
3384 | |
ICAM-2/CD102 Fc Chimera | |
Mouse myeloma cell line,NS0-derived human ICAM-2/CD102 protein, (N-terminus) Human ICAM-2 (Lys25-Gln223) Accession &Num; CAA33630 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |