missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPB6 Protein
A cDNA sequence encoding the HSPB6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12333-1mg
Additional Details : Weight : 0.01000kg
Specifications
HSPB6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEIPVP VQPSWLRRAS APLPGLSAPG RLFDQRFGEG LLEAELAALC PTTLAPYYLR APSVALPVAQ VPTDPGHFSV LLDVKHFSPE EIAVKVVGEH VEVHARHEER PDEHGFVARE FHRRYRLPPG VDPAAVTSAL SPEGVLSIQA APASAQAPPP AAAK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HSPB6 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 30% glycerol, 1mM DTT, 1mM EDTA and 0.1mM PMSF. |