missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HSP90 alpha Protein
Description
Reaction conditions will need to be optimized for each specific application. IMPORTANT: HSP90 alpha/HSP90AA1 works in conjunction with the co-chaperones such as p23/PTGES3--both proteins are typically required for enzymatic activity. For in vitro use we recommend an initial co-chaperone concentration of 2-3 µM, with an equimolar (or greater) concentration of HSP90 alpha/HSP90AA1.
Specifications
Specifications
| Accession Number | P07900.5 |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, KCl and TCEP. |
| Gene ID (Entrez) | 3320 |
| Molecular Weight (g/mol) | 85 kDa |
| Name | HSP90 alpha |
| Quantity | 50 μg |
| Source | E. coli-derived human HSP90 alpha protein Met1 - Asp732 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?