missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HSP70/HSPA1A Protein
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µg
Description
Reaction conditions will need to be optimized for each specific application. We recommend an initial HSP70/HSPA1A concentration of 2-3 μM for in vitro use. IMPORTANT: HSP40/DNAJB1 (Catalog # AP-110), or another suitable co-chaperone, is required for HSP70/HSPA1A activity and should be used at a concentration that is equimolar to HSP70/HSPA1A.
Specifications
Specifications
| Formulation | X mg/mL (X μM) in 50 mM HEPES pH 8, 100 mM NaCl, 5 mM DTT |
| Gene ID (Entrez) | 3303 |
| Molecular Weight (g/mol) | 70 kDa |
| Name | HSP70/HSPA1A |
| Quantity | 50 μg |
| Source | E. coli-derived human HSP70/HSPA1A protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Structural Form | Solution |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction