missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human HR6B/UBE2B His (N-Term) Protein

Product Code. 18413102 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413102 100 μg 100µg
18400772 10 μg 10µg
18423472 1 mg 1mg
18467251 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18413102 Supplier Novus Biologicals™ Supplier No. NBP235088100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) PAGE
Formulation A 0.2 um filtered concentrated solution in 50 mM HEPES, pH 7.6 with 125 mM NaCl, 10 % Glycerol, 1 mM DTT.
Gene ID (Entrez) 7320
Molecular Weight (g/mol) 19.0 kDa
Quantity 100 μg
Storage Requirements Store at -20C. Avoid freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of HR6B/UBE2B as determined by LAL method.
Gene Alias E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B
Gene Symbol UBE2B
Reconstitution Please contact technical support for detailed reconstitution instructions.
Research Category Biologically Active Proteins
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein HR6B/UBE2B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.