missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HPV B19 Capsid protein VP1 Protein

A DNA sequence encoding the Homo sapiens (Human) HPV B19 Capsid protein VP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7371-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048520

  • £384.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HPV B19 Capsid protein VP1 Protein
Research Use Only
Full Length
Human
His
Tris-based buffer, 50% glycerol
100 μg
Not determined.
Recombinant Protein
E. coli
MTSVNSAEAS TGAGGGGSNS VKSMWSEGAT FSANSVTCTF SRQFLIPYDP EHHYKVFSPA ASSCHNASGK EAKVCTISPI MGYSTPWRYL DFNALNLFFS PLEFQHLIEN YGSIAPDALT VTISEIAVKD VTDKTGGGVQ VTDSTTGRLC MLVDHEYKYP YVLGQGQDTL APELPIWVYF PPQYAYLTVG DVNTQGISGD SKKLASEESA FYVLEHSSFQ LLGTGGTASM SYKFPPVPPE NLEGCSQHFY EMYNPLYGSR LGVPDTLGGD PKFRSLTHED HAIQPQNFMP GPLVNSVSTK EGDSSNTGAG KALTGLSTGT SQNTRISLRP GPVSQPYHHW DTDKYVTGIN AISHGQTTYG NAEDKEYQQG VGRFPNEKEQ LKQLQGLNMH TYFPNKGTQQ YTDQIERPLM VGSVWNRRAL HYESQLWSKI PNLDDSFKTQ FAALGGWGLH QPPPQIFLKI LPQSGPIGGI KSMGITTLVQ YAVGIMTVTM TFKLGPRKAT GRWNPQPGVY PPHAAGHLPY VLYDPTATDA KQHHRHGYEK PEELWTAKSR VHPL
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers