missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HOMER2 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | HOMER2 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSHMMGEQP IFTTRAHVFQ IDPNTKKNWM PASKQAVTVS YFYDVTRNSY RIISVDGAKV IINSTITPNM TFTKTSQKFG QWADSRANTV FGLGFSSEQQ LTKFAEKFQE VKEAAKIAKD KTQEKIETSS NHSQASSVNG TDDEKASHAG PANTHLKSEN DKLKIALTQS AANVKKWEIE LQTLRESNAR LTTALQESAA SVEQWKRQFS ICRDENDRLR NKIDELEEQC SEINREKEKN TQLKRRIEEL EAELREKETE LKDLRKQSEI IPQLMSECEY VSEKLEAAER DNQNLEDKVR SLKTDIEESK YRQRHLKVEL KSFLEVLDGK IDDLHDFRRG LSKLGTDN |
| Buffer | The HOMER2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction