missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HOMER2 Protein
A cDNA sequence encoding the HOMER2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12292-10ug
Additional Details : Weight : 0.01000kg
Specifications
HOMER2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMMGEQP IFTTRAHVFQ IDPNTKKNWM PASKQAVTVS YFYDVTRNSY RIISVDGAKV IINSTITPNM TFTKTSQKFG QWADSRANTV FGLGFSSEQQ LTKFAEKFQE VKEAAKIAKD KTQEKIETSS NHSQASSVNG TDDEKASHAG PANTHLKSEN DKLKIALTQS AANVKKWEIE LQTLRESNAR LTTALQESAA SVEQWKRQFS ICRDENDRLR NKIDELEEQC SEINREKEKN TQLKRRIEEL EAELREKETE LKDLRKQSEI IPQLMSECEY VSEKLEAAER DNQNLEDKVR SLKTDIEESK YRQRHLKVEL KSFLEVLDGK IDDLHDFRRG LSKLGTDN | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HOMER2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |