missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein NANOG Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein NANOG, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8217-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001284627.1 | |
Homeobox protein NANOG Protein | |
Research Use Only | |
NANOG | |
Recombinant Protein | |
Yeast | |
MSVDPACPQS LPCFEASDCK ESSPMPVICG PEENYPSLQM SSAEMPHTET VSPLPSSMDL LIQDSPDSST SPKGKQPTSA EKSVAKKEDK VPVKKQKTRT VFSSTQLCVL NDRFQRQKYL SLQQMQELSN ILNLSYKQVK TWFQNQRMKS KRWQKNNWPK NSNGVTQKAS APTYPSLYSS YHQGCLVNPT GNLPMWSNQT WNNSTWSNQT QNIQSWSNHS WNTQTWCTQS WNNQAWNSPF YNCGEESLQS CMQFQPNSPA SDLEAALEAA GEGLNVIQQT TRYFSTPQTM DLFLNYSMNM QPEDV | |
Greater than 90% as determined by SDS-PAGE. |
79923 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |