missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Homeobox protein NANOG Protein

A DNA sequence encoding the Homo sapiens (Human) Homeobox protein NANOG, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8217-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043351

  • £1930.00 / 500µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001284627.1
Homeobox protein NANOG Protein
Research Use Only
NANOG
Recombinant Protein
Yeast
MSVDPACPQS LPCFEASDCK ESSPMPVICG PEENYPSLQM SSAEMPHTET VSPLPSSMDL LIQDSPDSST SPKGKQPTSA EKSVAKKEDK VPVKKQKTRT VFSSTQLCVL NDRFQRQKYL SLQQMQELSN ILNLSYKQVK TWFQNQRMKS KRWQKNNWPK NSNGVTQKAS APTYPSLYSS YHQGCLVNPT GNLPMWSNQT WNNSTWSNQT QNIQSWSNHS WNTQTWCTQS WNNQAWNSPF YNCGEESLQS CMQFQPNSPA SDLEAALEAA GEGLNVIQQT TRYFSTPQTM DLFLNYSMNM QPEDV
Greater than 90% as determined by SDS-PAGE.
79923
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers