missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Holliday junction recognition Protein

Product Code. 16071250
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16071250

Brand: enQuireBio™ QP6162ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Holliday junction recognition protein, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001269891.1
Gene ID (Entrez) 55355
Name Holliday junction recognition Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol HJURP
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MLGTLRAMEG EDVEDDQLLQ KLRASRRRFQ RRMQRLIEKY NQPFEDTPVV QMATLTYETP QGLRIWGGRL IKERNEGEIQ DSSMKPADRT DGSVQAAAWG PELPSHRTVL GADSKSGEVD ATSDQEESVA WALAPAVPQS PLKNELRRKY LTQVDILLQG AEYFECAGNR AGRDVRVTPL PSLASPAVPA PGYCSRISRK SPGDPAKPAS SPREWDPLHP SSTDMALVPR NDSLSLQETS SSSFLSSQPF EDDDICNVTI SDLYAGMLHS MSRLLSTKPS SIISTKTFIM QNWNSRRRHR YKSRMNKTYC KGARRSQRSS KENFIPCSEP VKGTGALRDC KNVLDVSCRK TGLKLEKAFL EVNRPQIHKL DPSWKERKVT PSKYSSLIYF DSSATYNLDE ENRFRTLKWL ISPVKIVSRP TIRQGHGENR QREIEIRFDQ LHREYCLSPR NQPRRMCLPD SWAMNMYRGG PASPGGLQGL ETRRLSLPSS KAKAKSLSEA FENLGKRSLE AGRCLPKSDS SSSLPKTNPT HSATRPQQTS DLHVQGNSSG IFRKSVSPSK TLSVPDKEVP GHGRNRYDEI KEEFDKLHQK YCLKSPGQMT VPLCIGVSTD KASMEVRYQT EGFLGKLNPD PHFQGFQKLP SSPLGCRKSL LGSTAIEAPS STCVARAITR DGTRDHQFPA KRPRLSEPQG SGRQGNSLGA SDGVDNTVRP GDQGSSSQPN SEERGENTSY RMEEKSDFML EKLETKSV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.