missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HN-1 / ARM2 Protein
A DNA sequence encoding the Homo sapiens (Human) HN-1 / ARM2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8233-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001002032.1 | |
HN-1 / ARM2 Protein | |
Research Use Only | |
HN1 | |
Recombinant Protein | |
Yeast | |
TTTTTFKGVD PNSRNSSRVL RPPGGGSNFS LGFDEPTEQP VRKNKMASNI FGTPEENQAS WAKSAGAKSS GGREDLESSG LQRRNSSEAS SGDFLDLKGE GDIHENVDTD LPGSLGQSEE KPVPAAPVPS PVAPAPVPSR RNPPGGKSSL VLG | |
Greater than 90% as determined by SDS-PAGE. |
51155 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |