missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HMGN1 Protein
Description
The Recombinant Human HMGN1 Protein is derived from E. coli. The Recombinant Human HMGN1 Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature. Upon receipt, store it immediately at the recommended storage temperatures.
Specifications
Specifications
| Quantity | 50 μg |
| Source | E. coli-derived human HMGN1 protein Pro2-Asp100 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Product Line | R&D Systems |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?