missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HMGB2 Protein

A cDNA sequence encoding the HMGB2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12282-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959928

  • £155.00 / 20µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HMGB2 Protein
Research Use Only
Recombinant Protein
Baculovirus
MGSSHHHHHH SSGLVPRGSH TGSMGKGDPN KPRGKMSSYA FFVQTCREEH KKKHPDSSVN FAEFSKKCSE RWKTMSAKEK SKFEDMAKSD KARYDREMKN YVPPKGDKKG KKKDPNAPKR PPSAFFLFCS EHRPKIKSEH PGLSIGDTAK KLGEMWSEQS AKDKQPYEQK AAKLKEKYEK DIAAYRAKGK SEAGKKGPGR PTGSKKKNEP EDEEEEEEEE DEDEEEEDED EE
Greater than 90% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HMGB2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 30% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers