missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLF Protein

A cDNA sequence encoding the HLF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12278-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939918

  • £1210.00 / 50µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HLF Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEKMSRPLPL NPTFIPPPYG VLRSLLENPL KLPLHHEDAF SKDKDKEKKL DDESNSPTVP QSAFLGPTLW DKTLPYDGDT FQLEYMDLEE FLSENGIPPS PSQHDHSPHP PGLQPASSAA PSVMDLSSRA SAPLHPGIPS PNCMQSPIRP GQLLPANRNT PSPIDPDTIQ VPVGYEPDPA DLALSSIPGQ EMFDPRKRKF SEEELKPQPM IKKARKVFIP DDLKDDKYWA RRRKNNMAAK RSRDARRLKE NQIAIRASFL EKENSALRQE VADLRKELGK CKNILAKYEA RHGPL
Greater than 85.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HLF protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.2M NaCl and 0.1mM PMSF.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers