missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histidine--tRNA ligase, cytoplasmic Protein

Product Code. 15912798
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
This item is not returnable. View return policy

Product Code. 15912798

Brand: enQuireBio™ QP10038ba20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Histidine--tRNA ligase, cytoplasmic, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001244969.1
Gene ID (Entrez) 3035
Name Histidine--tRNA ligase, cytoplasmic Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol HARS
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence AERAALEELV KLQGERVRGL KQQKASAELI EEEVAKLLKL KAQLGPDESK QKFVLKTPKG TRDYSPRQMA VREKVFDVII RCFKRHGAEV IDTPVFELKE TLMGKYGEDS KLIYDLKDQG GELLSLRYDL TVPFARYLAM NKLTNIKRYH IAKVYRRDNP AMTRGRYREF YQCDFDIAGN FDPMIPDAEC LKIMCEILSS LQIGDFLVKV NDRRILDGMF AICGVSDSKF RTICSSVDKL DKVSWEEVKN EMVGEKGLAP EVADRIGDYV QQHGGVSLVE QLLQDPKLSQ NKQALEGLGD LKLLFEYLTL FGIDDKISFD LSLARGLDYY TGVIYEAVLL QTPAQAGEEP LGVGSVAAGG RYDGLVGMFD PKGRKVPCVG LSIGVERIFS IVEQRLEALE EKIRTTETQV LVASAQKKLL EERLKLVSEL WDAGIKAELL YKKNPKLLNQ LQYCEEAGIP LVAIIGEQEL KDGVIKLRSV TSREEVDVRR EDLVEEIKRR TGQ
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.