missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human His6-USP30 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Reaction conditions will need to be optimized for each specific application. We recommend an initial USP30 concentration of 20-100 nM in reactions utilizing Ubiquitin-AMC or Ubiquitin-Rhodamine substrates (U-550 or U-555), or 200-500 nM when digesting recombinant Polyubiquitin chain substrates.
Specifications
Specifications
| Accession Number | Q70CQ3.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, Glycerol and DTT. |
| Gene ID (Entrez) | 84749 |
| Molecular Weight (g/mol) | M.W. (theoretical): 54 kDa |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP30 protein Thr57 - Glu517 with a N-terminal Met and a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Alias | Deubiquitinating enzyme 30, EC 3.1.2.15, EC 3.4.19.12, FLJ40511, MGC10702, ubiquitin carboxyl-terminal hydrolase 30, ubiquitin specific peptidase 30, ubiquitin specific protease 30, ubiquitin thioesterase 30, Ubiquitin thiolesterase 30, ubiquitin-specific protease 30, Ubiquitin-specific-processing protease 30, Ub-specific protease 30 |
| Common Name | USP30 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction