missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HGF Protein
Click to view available options
Quantity:
25 μg
250 μg
5 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl.
Unit Size:
250µg
25µg
5µg
Description
The Recombinant Human HGF Protein is derived from Sf 21 (baculovirus). The Recombinant Human HGF Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P14210.2 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 3082 |
| Molecular Weight (g/mol) | Predicted MW: 53.7kDa ( alpha chain), 26kDa ( beta chain) Observed MW: 60-70kDa, non-reducing conditions 60kDa, and 33-34kDa, reducing conditions |
| Name | HGF |
| Quantity | 250 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human HGF protein Gln32-Ser728 (Asp384Asn, Asp416Asn, Leu622Ser, His645Arg, Val678Ile) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Gene Alias | deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction