missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HGF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 294-HG-025
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human HGF Protein is derived from Sf 21 (baculovirus). The Recombinant Human HGF Protein has been validated for the following applications: Bioactivity.Specifications
P14210.2 | |
3082 | |
HGF | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human HGF protein Gln32-Ser728 (Asp384Asn, Asp416Asn, Leu622Ser, His645Arg, Val678Ile) | |
Disulfide-linked heterodimer | |
Unconjugated | |
Reconstitute at 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. |
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein. | |
Predicted MW: 53.7kDa ( alpha chain), 26kDa ( beta chain) Observed MW: 60-70kDa, non-reducing conditions 60kDa, and 33-34kDa, reducing conditions | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
Recombinant | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |