missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HGF (NS0-expressed) Protein
Click to view available options
Quantity:
100 μg
250 μg
Unit Size:
100µg
250µg
Description
The Recombinant Human HGF (NS0-expressed) Protein is derived from NS0. The Recombinant Human HGF (NS0-expressed) Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Accession Number | P14210.2 |
| For Use With (Application) | Binding Activity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 3082 |
| Molecular Weight (g/mol) | alpha chain: 53.7 kDa beta chain: 26 kDa |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line, NS0-derived human HGF protein Gln32-Ser728 (Asp384Asn, Asp416Asn, Leu622Ser, His645Arg, Val678Ile) |
| Storage Requirements | Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction