missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Hexokinase 2 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to phosphorylate glucose. The specific activity is >1,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P52789 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, DTT, Glucose and Glycerol. |
| Gene ID (Entrez) | 3099 |
| Molecular Weight (g/mol) | M.W. (Observed): 100-105 kDa, reducing conditions; M.W. (theoretical): 102 kDa |
| Name | Hexokinase 2 |
| Quantity | 20 μg |
| Source | E. coli-derived human Hexokinase 2 protein Phe11-Arg917, with N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction