missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein 1

A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein K, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8415-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024491

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001305115.1
Heterogeneous nuclear ribonucleoprotein 1
Research Use Only
HNRPK
Recombinant Protein
E. coli
TEQPEETFPN TETNGEFGKR PAEDMEEEQA FKRSRNTDEM VELRILLQSK NAGAVIGKGG KNIKALRTDY NASVSVPDSS GPERILSISA DIETIGEILK KIIPTLEEGL QLPSPTATSQ LPLESDAVEC LNYQHYKGSD FDCELRLLIH QSLAGGIIGV KGAKIKELRE NTQTTIKLFQ ECCPHSTDRV VLIGGKPDRV VECIKIILDL ISESPIKGRA QPYDPNFYDE TYDYGGFTMM FDDRRGRPVG FPMRGRGGFD RMPPGRGGRP MPPSRRDYDD MSPRRGPPPP PPGRGGRGGS RARNLPLPPP PPPRGGDLMA YDRRGRPGDR YDGMVGFSAD ETWDSAIDTW SPSEWQMAYE PQGGSGYDYS YAGGRGSYGD LGGPIITTQV TIPKDLAGSI IGKGGQRIKQ IRHESGASIK IDEPLEGSED RIITITGTQD QIQNAQYLLQ NSVKQYS
Greater than 90% as determined by SDS-PAGE.
3190
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers