missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human HERPUD1 His Protein

Product Code. 18214290 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
0.25 mg
Unit Size:
0.05mg
0.25mg
This item is not returnable. View return policy

Product Code. 18214290

Brand: Novus Biologicals™ NBP2518620.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-263 of Human HERPUD1 The Recombinant Human HERPUD1 Protein is derived from E. coli. The Recombinant Human HERPUD1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffer saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 9709
Molecular Weight (g/mol) TMW: 31.6kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias HERPMIF1, homocysteine-inducible endoplasmic reticulum stress-inducible ubiquitin-likedomain member 1 protein, homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-likedomain member 1, homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domainmember 1 protein, KIAA0025MMS-inducible, Methyl methanesulfonate (MMF)-inducible fragment protein 1, Mif1, SUP
Common Name HERPUD1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.