missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HEBP1 Protein

A cDNA sequence encoding the HEBP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12214-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939718

  • £99.15 / 2µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HEBP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLGMIKN SLFGSVETWP WQVLSKGDKE EVAYEERACE GGKFATVEVT DKPVDEALRE AMPKVAKYAG GTNDKGIGMG MTVPISFAVF PNEDGSLQKK LKVWFRIPNQ FQSDPPAPSD KSVKIEEREG ITVYSMQFGG YAKEADYVAQ ATRLRAALEG TATYRGDIYF CTGYDPPMKP YGRRNEIWLL KT
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HEBP1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers