missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heat shock protein 75 kDa, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Heat shock protein 75 kDa, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8452-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001258978.1 | |
Heat shock protein 75 kDa, mitochondrial Protein | |
Research Use Only | |
OTUB1 | |
Recombinant Protein | |
E. coli | |
STQTAEDKEE PLHSIISSTE SVQGSTSKHE FQAETKKLLD IVARSLYSEK EVFIRELISN ASDALEKLRH KLVSDGQALP EMEIHLQTNA EKGTITIQDT GIGMTQEELV SNLGTIARSG SKAFLDALQN QAEASSKIIG QFGVGFYSAF MVADRVEVYS RSAAPGSLGY QWLSDGSGVF EIAEASGVRT GTKIIIHLKS DCKEFSSEAR VRDVVTKYSN FVSFPLYLNG RRMNTLQAIW MMDPKDVRE | |
Greater than 90% as determined by SDS-PAGE. |
10131 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |