missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heat shock 70 kDa protein 1

Product Code. 16002421
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16002421

Brand: enQuireBio™ QP8058ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Heat shock 70 kDa protein 6, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_002146.2
Gene ID (Entrez) 3310
Name Heat shock 70 kDa protein 1
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol HSPA6
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MQAPRELAVG IDLGTTYSCV GVFQQGRVEI LANDQGNRTT PSYVAFTDTE RLVGDAAKSQ AALNPHNTVF DAKRLIGRKF ADTTVQSDMK HWPFRVVSEG GKPKVRVCYR GEDKTFYPEE ISSMVLSKMK ETAEAYLGQP VKHAVITVPA YFNDSQRQAT KDAGAIAGLN VLRIINEPTA AAIAYGLDRR GAGERNVLIF DLGGGTFDVS VLSIDAGVFE VKATAGDTHL GGEDFDNRLV NHFMEEFRRK HGKDLSGNKR ALRRLRTACE RAKRTLSSST QATLEIDSLF EGVDFYTSIT RARFEELCSD LFRSTLEPVE KALRDAKLDK AQIHDVVLVG GSTRIPKVQK LLQDFFNGKE LNKSINPDEA VAYGAAVQAA VLMGDKCEKV QDLLLLDVAP LSLGLETAGG VMTTLIQRNA TIPTKQTQTF TTYSDNQPGV FIQVYEGERA MTKDNNLLGR FELSGIPPAP RGVPQIEVTF DIDANGILSV TATDRSTGKA NKITITNDKG RLSKEEVERM VHEAEQYKAE DEAQRDRVAA KNSLEAHVFH VKGSLQEESL RDKIPEEDRR KMQDKCREVL AWLEHNQLAE KEEYEHQKRE LEQICRPIFS RLYGGPGVPG GSSCGTQARQ GDPSTGP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.