missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDDC2 Protein
A cDNA sequence encoding the HDDC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12204-20ug
Additional Details : Weight : 0.01000kg
Specifications
HDDC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASVSSA TFSGHGARSL LQFLRLVGQL KRVPRTGWVY RNVQRPESVS DHMYRMAVMA MVIKDDRLNK DRCVRLALVH DMAECIVGDI APADNIPKEE KHRREEEAMK QITQLLPEDL RKELYELWEE YETQSSAEAK FVKQLDQCEM ILQASEYEDL EHKPGRLQDF YDSTAGKFNH PEIVQLVSEL EAERSTNIAA AASEPHS | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HDDC2 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |