missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HCP1 Protein
Shop All Bio Techne Products
Click to view available options
:
10 μg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Immunoaffinity Purification |
| Formulation | 25 mM Tris-HCl pH 8.0 in 2% glycerol. |
| Gene ID (Entrez) | 113235 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 47.5 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Avoid freeze/thaw cycles. |
| Gene Alias | FLJ39875, G21, HCP1PCFT/HCP1, Heme carrier protein 1, MGC9564, PCFTsolute carrier family 46, member 1, proton-coupled folate transporter, solute carrier family 46 (folate transporter), member 1, Solute carrier family 46 member 1 |
| Gene Symbol | SLC46A1 |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | HCP1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction