missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HBEGF / DTR Protein
Recombinant Protein
Brand: enQuireBio™ QP5305-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001936.1 | |
HBEGF / DTR Protein | |
Research Use Only | |
HBEGF | |
Recombinant Protein | |
E. coli | |
MDLQEADLDL . LRVTLSSKP Q. ALATPNKE EH. GKRKKKG KGL. GKKRDP CLRK. YKDFC IHGEC. KYVK ELRAPS. CIC HPGYHGERCH GLSL | |
Greater than 95% as determined by SDS-PAGE. |
1839 | |
50 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
3T3 cell proliferation is significantly induced by less than 1 ng/ml Human HBEGF / DTR, (activity greater than 1.0 x 10^6 units/mg). | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |