missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HBA2 Protein

A cDNA sequence encoding the HBA2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12118-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939428

  • £2975.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HBA2 Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMVL SPADKTNVKA AWGKVGAHAG EYGAEALERM FLSFPTTKTY FPHFDLSHGS AQVKGHGKKV ADALTNAVAH VDDMPNALSA LSDLHAHKLR VDPVNFKLLS HCLLVTLAAH LPAEFTPAVH ASLDKFLASV STVLTSKYR
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HBA2 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 2mM DTT, 2M urea and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers