missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HAUS1 Protein
A cDNA sequence encoding the HAUS1 was constructed and used to recombinantly synthesize the protein.
£104.00 - £6030.00
Specifications
Name | HAUS1 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His |
Host Species | E. coli |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969388
|
enQuireBio™
QP12106-2UG |
2 μg |
£104.00
2µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15949388
|
enQuireBio™
QP12106-10UG |
10 μg |
£159.00
10µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15959388
|
enQuireBio™
QP12106-1MG |
1 mg |
£6030.00
1mg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
HAUS1 Protein | |
His | |
Human | |
Greater than 85.0% as determined by SDS-PAGE. |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEPQEE RETQVAAWLK KIFGDHPIPQ YEVNPRTTEI LHHLSERNRV RDRDVYLVIE DLKQKASEYE SEAKYLQDLL MESVNFSPAN LSSTGSRYLN ALVDSAVALE TKDTSLASFI PAVNDLTSDL FRTKSKSEEI KIELEKLEKN LTATLVLEKC LQEDVKKAEL HLSTERAKVD NRRQNMDFLK AKSEEFRFGI KAAEEQLSAR GMDASLSHQS LVALSEKLAR LKQQTIPLKK KLESYLDLMP NPSLAQVKIE EAKRELDSIE AELTRRVDMM EL |