missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Haptoglobin Protein

Product Code. 15959378
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15959378

Brand: enQuireBio™ QP12103HIS1mg

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A cDNA sequence encoding the Haptoglobin was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 3240
Name Haptoglobin Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol HP
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence MSALGAVIAL LLWGQLFAVD SGNDVTDIAD DGCPKPPEIA HGYVEHSVRY QCKNYYKLRT EGDGVYTLND KKQWINKAVG DKLPECEADD GCPKPPEIAH GYVEHSVRYQ CKNYYKLRTE GDGVYTLNNE KQWINKAVGD KLPECEAVCG KPKNPANPVQ RILGGHLDAK GSFPWQAKMV SHHNLTTGAT LINEQWLLTT AKNLFLNHSE NATAKDIAPT LTLYVGKKQL VEIEKVVLHP NYSQVDIGLI KLKQKVSVNE RVMPICLPSK DYAEVGRVGY VSGWGRNANF KFTDHLKYVM LPVADQDQCI RHYEGSTVPE KKTPKSPVGV QPILNEHTFC AGMSKYQEDT CYGDAGSAFA VHDLEEDTWY ATGILSFDKS CAVAEYGVYV KVTSIQDWVQ KTIAEN
Buffer Lyophilized with no additives.
Purity or Quality Grade Greater than 95.0%.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.