missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GZMK Protein
A cDNA sequence encoding the GZMK was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12073-HIS-5ug
This item is not returnable.
View return policy
Specifications
3003 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
ADPIIGGKEV SPHSRPFMAS IQYGGHHVCG GVLIDPQWVL TAAHCQYRFT KGQSPTVVLG AHSLSKNEAS KQTLEIKKFI PFSRVTSDPQ SNDIMLVKLQ TAAKLNKHVK MLHIRSKTSL RSGTKCKVTG WGATDPDSLR PSDTLREVTV TVLSRKLCNS QSYYNGDPFI TKDMVCAGDA KGQKDSCKGD SGGPLICKGV FHAIVSGGHE CGVATKPGIY TLLTKKYQTW IKSNLVPPHT NHHHHHH | |
Greater than 95.0% as determined by analysis by SDS-PAGE. |
GZMK Protein | |
Research Use Only | |
GZMK | |
Human | |
His | |
GZMK protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 20% glycerol and 1mM DTT. |