missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Guanine deaminase Protein

Product Code. 18193918 Shop All Bio Techne Products
Click to view available options
Quantity:
20 μg
5 μg
Unit Size:
20µg
5µg
This item is not returnable. View return policy

Product Code. 18193918

Brand: Novus Biologicals™ NBP2496925UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-454 of Human Guanine deaminase The Recombinant Human Guanine deaminase Protein is derived from E. coli. The Recombinant Human Guanine deaminase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol, 1mM DTT
Gene ID (Entrez) 9615
Molecular Weight (g/mol) 51 kDa
Name Recombinant Human Guanine deaminase Protein
Purification Method >90%, by SDS-PAGE
Quantity 5 μg
Source E.coli
Storage Requirements −80°C in dark, avoid freeze-thaw cycles
Regulatory Status RUO
Gene Alias CYPIN, EC 3.5.4.3, GAH, GUANASE, Guanine aminase, Guanine aminohydrolase, guanine deaminase, KIAA1258, MGC9982, NEDASIN, p51-nedasin
Gene Symbol GDA
Product Type Recombinant protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.