missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GTP-binding protein SAR1a Protein
A DNA sequence encoding the Homo sapiens (Human) GTP-binding protein SAR1a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8119-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001136120.1 | |
GTP-binding protein SAR1a Protein | |
Research Use Only | |
SAR1A | |
Recombinant Protein | |
E. coli | |
MSFIFEWIYN GFSSVLQFLG LYKKSGKLVF LGLDNAGKTT LLHMLKDDRL GQHVPTLHPT SEELTIAGMT FTTFDLGGHE QARRVWKNYL PAINGIVFLV DCADHSRLVE SKVELNALMT DETISNVPIL ILGNKIDRTD AISEEKLREI FGLYGQTTGK GNVTLKELNA RPMEVFMCSV LKRQGYGEGF RWLSQYID | |
Greater than 80% as determined by SDS-PAGE. |
56681 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |