missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GTP-binding protein SAR1a Protein

A DNA sequence encoding the Homo sapiens (Human) GTP-binding protein SAR1a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8119-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002771

  • £1735.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001136120.1
GTP-binding protein SAR1a Protein
Research Use Only
SAR1A
Recombinant Protein
E. coli
MSFIFEWIYN GFSSVLQFLG LYKKSGKLVF LGLDNAGKTT LLHMLKDDRL GQHVPTLHPT SEELTIAGMT FTTFDLGGHE QARRVWKNYL PAINGIVFLV DCADHSRLVE SKVELNALMT DETISNVPIL ILGNKIDRTD AISEEKLREI FGLYGQTTGK GNVTLKELNA RPMEVFMCSV LKRQGYGEGF RWLSQYID
Greater than 80% as determined by SDS-PAGE.
56681
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers