missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GTP-binding nuclear protein Ran Protein

A DNA sequence encoding the Homo sapiens (Human) GTP-binding nuclear protein Ran, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6583-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084320

  • £384.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001287725.1
GTP-binding nuclear protein Ran Protein
Research Use Only
RAN
Recombinant Protein
E. coli
AAQGEPQVQF KLVLVGDGGT GKTTFVKRHL TGEFEKKYVA TLGVEVHPLV FHTNRGPIKF NVWDTAGQEK FGGLRDGYYI QAQCAIIMFD VTSRVTYKNV PNWHRDLVRV CENIPIVLCG NKVDIKDRKV KAKSIVFHRK KNLQYYDISA KSNYNFEKPF LWLARKLIGD PNLEFVAMPA LAPPEVVMDP ALAAQYEHDL EVAQTTALPD EDDDL
Greater than 80% as determined by SDS-PAGE.
5901
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers