missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GTF3C6 Protein
A cDNA sequence encoding the GTF3C6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12067-2ug
Additional Details : Weight : 0.01000kg
Specifications
GTF3C6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAAADE RSPEDGEDEE EEEQLVLVEL SGIIDSDFLS KCENKCKVLG IDTERPILQV DSCVFAGEYE DTLGTCVIFE ENVEHADTEG NNKTVLKYKC HTMKKLSMTR TLLTEKKEGE ENIGGVEWLQ IKDNDFSYRP NMICNFLHEN EDEEVVASAP DKSLELEEEE IQMNDSSNLS CEQEKPMHLE IEDSGPLIDI PSETEGSVFM ETQMLP | |
Greater than 85.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GTF3C6 protein solution (0.5 mg/ml) containing PBS buffer (pH 7.4), 10% glycerol and 1mM DTT. |