missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human GRP78/HSPA5 His Protein

Product Code. 18355072 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
Unit Size:
100µg
200µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355072 100 μg 100µg
18374302 50 μg 50µg
18320254 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18355072 Supplier Novus Biologicals™ Supplier No. NBP318319100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

The protein has ATPase activity at the time of manufacture of 2.3uM phosphate liberated/hr/ug protein in a 200ul reaction at 37C (pH 8) in the presence of 20ul of 1mM ATP using a Malachite Green assay.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity, ELISA, Functional Assay, SDS-PAGE, Western Blot
Formulation 20mM Tris/HCl pH7.5, 50mM KCl, 5mM MgCl2, 1mM DTT
Gene ID (Entrez) 3309
Molecular Weight (g/mol) M.W. Theoretical: 78 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2
Gene Symbol HSPA5
Research Category Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR)
Purity or Quality Grade >85%
Protein GRP78/HSPA5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.