missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Growth Hormone Protein

Product Code. 18402102 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18402102 500 μg 500µg
18490422 1 mg 1mg
18471402 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18402102 Supplier R&D Systems™ Supplier No. NBP234890500UG
  • ONLINE EXCLUSIVE PRICE
    £647.76
    £515.55 / 500µg
    Save £132.21
    20% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

Growth Hormone protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using rat Nb2-11 lymphoma cells is less than 0.1 ng/mL, corresponding to a specific activity of >1.0 x 107 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01241
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution with 20mM PB, pH 7.0.
Gene ID (Entrez) 2688
Molecular Weight (g/mol) 22 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of Growth Hormone as determined by LAL method.
Gene Alias GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N
Gene Symbol GH1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins
Product Line Novus
Purity or Quality Grade >98% pure by SDS-PAGE and HPLC
Protein Growth Hormone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.