missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Growth Hormone (GH) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£183.58
Specifications
Gene ID (Entrez) | 2688 |
---|---|
Molecular Weight (g/mol) | 22 kDa |
Name | Growth Hormone |
Quantity | 25μg |
Source | E. coli-derived human Growth Hormone protein Phe27-Phe217,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18176500
|
R&D Systems™
1067-GH-025/CF |
Lyophilized from a 0.2μm filtered solution in Tris and NaCl. |
£183.58
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18118040
|
R&D Systems™
1067-GH-025 |
Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein. |
£183.58
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human Growth Hormone (GH) Protein is derived from E. coli. The Recombinant Human Growth Hormone (GH) Protein has been validated for the following applications: Bioactivity.Specifications
2688 | |
Growth Hormone | |
E. coli-derived human Growth Hormone protein Phe27-Phe217,with an N-terminal Met | |
GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin | |
Recombinant |
22 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |