missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Novus Biologicals™ Recombinant Human Granzyme M GST (N-Term) Protein
Handla allt Bio Techne Produkter
Klicka för att se tillgängliga alternativ
Quantity:
10 μg
25 μg
Förpackningsstorlek:
10µg
25µg
Beskrivning
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifikationer
Specifikationer
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 3004 |
| Molecular Weight (g/mol) | 37.73 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | EC 3.4.21, EC 3.4.21.-, granzyme M, granzyme M (lymphocyte met-ase 1), Hu-Met-1, LMET1, lymphocyte met-ase 1, Met-1 serine protease, MET1met-ase, Met-ase, Natural killer cell granular protease |
| Gene Symbol | GZMM |
| Research Category | Adaptive Immunity, Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering