missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GPX7 Protein

A cDNA sequence encoding the GPX7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12045-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959188

  • £2975.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GPX7 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMQQEQD FYDFKAVNIR GKLVSLEKYR GSVSLVVNVA SECGFTDQHY RALQQLQRDL GPHHFNVLAF PCNQFGQQEP DSNKEIESFA RRTYSVSFPM FSKIAVTGTG AHPAFKYLAQ TSGKEPTWNF WKYLVAPDGK VVGAWDPTVS VEEVRPQITA LVRKLILLKR EDL
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GPX7 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers