missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GPX3 Protein

A cDNA sequence encoding the GPX3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12044-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939188

  • £5730.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GPX3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQSRGQEKSK MDCHGGISGT IYEYGALTID GEEYIPFKQY AGKYVLFVNV ASYCGLTGQY IELNALQEEL APFGLVILGF PCNQFGKQEP GENSEILPTL KYVRPGGGFV PNFQLFEKGD VNGEKEQKFY TFLKNSCPPT SELLGTSDRL FWEPMKVHDI RWNFEKFLVG PDGIPIMRWH HRTTVSNVKM DILSYMRRQA ALGVKRK
Greater than 85% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GPX3 solution contains 20mM Tris-HCl buffer (pH 7.5), 40% glycerol, 0.15M NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers