missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human GPT2 His Protein
Description
Specific activity is > 100units/mg, and is defined as the amount of enzyme that cleaves 1 umole of L-Alanine to L-Glutamate per minute at pH 7.5 at 37C
Specifications
Specifications
| Concentration | 0.5 mg/ml |
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 7.5), 30% glycerol, 2mM DTT, 0.2M NaCl |
| Gene ID (Entrez) | 84706 |
| Molecular Weight (g/mol) | M.W. Theoretical: 60.3 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | AAT2, alanine aminotransferase 2, ALT2EC 2.6.1.2, Glutamate pyruvate transaminase 2, glutamic pyruvate transaminase (alanine aminotransferase) 2, Glutamic--alanine transaminase 2, glutamic-pyruvate transaminase 2, Glutamic--pyruvic transaminase 2, GPT 2 |
| Gene Symbol | GPT2 |
| Research Category | metabolism |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?