missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human GPR63 Protein
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantity:
10 μg
Dimensione della confezione:
10µg
Descrizione
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifica
Specifica
| For Use With (Application) | Immunoaffinity Purification |
| Formulation | 25 mM Tris-HCl pH 8.0 in 2% glycerol. |
| Gene ID (Entrez) | 81491 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 47.5 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Avoid freeze/thaw cycles. |
| Gene Alias | brain expressed G-protein-coupled receptor PSP24 beta, G protein-coupled receptor 63, probable G-protein coupled receptor 63, PSP24-2, PSP24-beta, PSP24BPSP24(beta) |
| Gene Symbol | GPR63 |
| Research Category | GPCR |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto